accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3FFFFFF

CAS RN 309-88-6

IUPAC Name: diethyl(1,1,2,3,3,3-hexafluoropropyl)amine
Synonyms: diethyl(1,1,2,3,3,3-hexafluoropropyl)amine
Molecular Weight (g/mol): 223.16
Molecular Formula: C7H11F6N
InChi Key: BNTFCVMJHBNJAR-UHFFFAOYNA-N
SMILES: CCN(CC)C(F)(F)C(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)