accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30925-07-6

ClClNH2SS(R)NH2HOOOHOHH

CAS RN 30925-07-6

IUPAC Name: dihydrogen (2R)-2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dichloride
Synonyms: dihydrogen (2R)-2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dichloride
Molecular Weight (g/mol): 313.21
Molecular Formula: C6H14Cl2N2O4S2
InChi Key: HHGZUQPEIHGQST-VBCXBIGUSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC(CSSC[C@H](N)C(O)=O)C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)