accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNOOOHO

CAS RN 3095-38-3

IUPAC Name: 3,5-dimethyl-4-nitrobenzoic acid
Synonyms: 3,5-dimethyl-4-nitrobenzoic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: RBAVFNOGEPCOQI-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results