accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3095-94-1

H3CPClClO

CAS RN 3095-94-1

IUPAC Name: octylphosphonoyl dichloride
Synonyms: octylphosphonoyl dichloride
Molecular Weight (g/mol): 231.1
Molecular Formula: C8H17Cl2OP
InChi Key: RTHCRCDXILKTCY-UHFFFAOYSA-N
SMILES: CCCCCCCCP(Cl)(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)