CAS RN 30951-66-7
IUPAC Name:
2-(3-bromophenyl)propan-2-ol
Synonyms:
2-(3-bromophenyl)propan-2-ol
Molecular Weight (g/mol):
215.09
Molecular Formula:
C9H11BrO
InChi Key:
ZRFMJMFYMQAUDO-UHFFFAOYSA-N
SMILES:
CC(C)(O)C1=CC(Br)=CC=C1
1
–
1
of
1
results
1
2-(3-Bromophenyl)propan-2-ol, 97%, Thermo Scientific™
CAS: 30951-66-7 Molecular Formula: C9H11BrO Molecular Weight (g/mol): 215.09 InChI Key: ZRFMJMFYMQAUDO-UHFFFAOYSA-N Synonym: 2-3-bromophenyl propan-2-ol,2-3-bromophenyl-2-propanol,acmc-209hjh,2-3-bromophenyl-propan-2-ol,2-3-bromo-phenyl-propan-2-ol,3-alpha-hydroxyisopropyl-1-bromobenzene PubChem CID: 15072383 IUPAC Name: 2-(3-bromophenyl)propan-2-ol SMILES: CC(C)(C1=CC(=CC=C1)Br)O