accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30980-84-8

H2OOIrO

CAS RN 30980-84-8

IUPAC Name: dioxoiridium hydrate
Synonyms: dioxoiridium hydrate
Molecular Weight (g/mol): 242.23
Molecular Formula: H2IrO3
InChi Key: YEPXCFHWKVNVAX-UHFFFAOYSA-N
SMILES: O.O=[Ir]=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results