accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNH2NHO

CAS RN 30991-42-5

IUPAC Name: 2-hydroxy-3-methylbenzohydrazide
Synonyms: 2-hydroxy-3-methylbenzohydrazide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: UWNTVIWGDXKENJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(=O)NN)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results