accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31011-57-1

IUPAC Name: bis(oxolane); tetrachlorotitanium
Synonyms: bis(THF); titanium tetrachloride
Molecular Weight (g/mol): 333.88
Molecular Formula: C8H16Cl4O2Ti
InChi Key: LXWBMENBONGPSB-UHFFFAOYSA-J
SMILES: Cl[Ti](Cl)(Cl)Cl.C1CCOC1.C1CCOC1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results