accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(R)OH(S)HOHNCH3O(E)

CAS RN 3102-57-6

IUPAC Name: N-[(2S,3R,4E)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
Synonyms: C2-ceramide N-acetylsphingosine N-(acetyl)-ceramide
Molecular Weight (g/mol): 341.54
Molecular Formula: C20H39NO3
InChi Key: BLTCBVOJNNKFKC-QUDYQQOWSA-N
SMILES: CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Product Type 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results