accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31027-31-3

IUPAC Name: 1-isocyanato-4-(propan-2-yl)benzene
Synonyms: 1-isocyanato-4-isopropylbenzene
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: PJVRNNRZWASOIT-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)N=C=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results