accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3105-55-3

NaOOOO(Z)

CAS RN 3105-55-3

IUPAC Name: sodium (2Z)-but-2-enedioate
Synonyms: sodium maleate
Molecular Weight (g/mol): 137.05
Molecular Formula: C4H2NaO4
InChi Key: VRVKOZSIJXBAJG-ODZAUARKSA-L
SMILES: [Na+].[O-]C(=O)\C=C/C([O-])=O
Quantity 
  • (4)
  • (4)
pH 
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (8)
  • (8)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)