accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3107-98-0

IUPAC Name: 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate
Synonyms: 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate
Molecular Weight (g/mol): 318.12
Molecular Formula: C8H6F8O4
InChi Key: XPXVIIILXUOEQA-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results