accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrN

CAS RN 31106-82-8

IUPAC Name: 2-(bromomethyl)pyridine
Synonyms: 2-(bromomethyl)pyridine
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: OFPWMRMIFDHXFE-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC=N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)