accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNHClClCl

CAS RN 31118-87-3

IUPAC Name: (2,4,6-trichlorophenyl)thiourea
Synonyms: 2,4,6-trichlorophenylthiourea
Molecular Weight (g/mol): 255.54
Molecular Formula: C7H5Cl3N2S
InChi Key: ZUSZPZPGZMEXLA-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Cl)C=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results