accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31126-95-1

IUPAC Name: tetrakis(trifluoroacetic acid) dirhodium
Synonyms: tetrakis(trifluoroacetic acid) dirhodium
Molecular Weight (g/mol): 661.9
Molecular Formula: C8H4F12O8Rh2
InChi Key: SZQVMUPTZFMHQT-UHFFFAOYSA-N
SMILES: [Rh].[Rh].OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results