accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31166-44-6

IUPAC Name: 4-[(benzyloxy)carbonyl]piperazin-1-ium
Synonyms: 4-[(benzyloxy)carbonyl]piperazin-1-ium
Molecular Weight (g/mol): 221.28
Molecular Formula: C12H17N2O2
InChi Key: CTOUWUYDDUSBQE-UHFFFAOYSA-O
SMILES: O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)