accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOSOOFFFFOFFFFFFOFFFFFFF

CAS RN 31175-20-9

IUPAC Name: 1,1,2,2-tetrafluoro-2-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)ethane-1-sulfonic acid; tetrafluoroethene
Synonyms: 1,1,2,2-tetrafluoro-2-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)ethanesulfonic acid; tetrafluoroethylene
Molecular Weight (g/mol): 544.13
Molecular Formula: C9HF17O5S
InChi Key: FOYUGSIADQEOEK-UHFFFAOYNA-N
SMILES: FC(F)=C(F)F.OS(=O)(=O)C(F)(F)C(F)(F)OC(F)(C(F)(F)F)C(F)(F)OC(F)=C(F)F
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (20)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)