accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3121-61-7

IUPAC Name: 2-methoxyethyl prop-2-enoate
Synonyms: methoxyethyl acrylate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: HFCUBKYHMMPGBY-UHFFFAOYSA-N
SMILES: COCCOC(=O)C=C
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results