accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNHNOOH

CAS RN 312496-16-5

IUPAC Name: 2-(pyridin-3-yl)-1H-1,3-benzodiazole-6-carboxylic acid
Synonyms: 2-(pyridin-3-yl)-3H-1,3-benzodiazole-5-carboxylic acid
Molecular Weight (g/mol): 239.23
Molecular Formula: C13H9N3O2
InChi Key: DHRPCXYIVJXXJR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2N=C(NC2=C1)C1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results