accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 312623-79-3

IUPAC Name: 2-(4-methoxyphenyl)-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-ol
Synonyms: 2-(4-methoxyphenyl)-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-ol
Molecular Weight (g/mol): 264.28
Molecular Formula: C14H16O5
InChi Key: SSKZQXMNNREVNP-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C1OCC2OC=CC(O)C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results