accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH2FClZn

CAS RN 312693-07-5

IUPAC Name: (4-fluorophenyl)methanide; chlorozincylium
Synonyms: (4-fluorophenyl)methanide; chlorozincylium
Molecular Weight (g/mol): 209.95
Molecular Formula: C7H6ClFZn
InChi Key: HEXMIUITECMJJV-UHFFFAOYSA-M
SMILES: Cl[Zn+].[CH2-]C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results