accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOI

CAS RN 31295-66-6

IUPAC Name: 4-iodo-5-methyl-3-phenyl-1,2-oxazole
Synonyms: 4-iodo-5-methyl-3-phenyl-1,2-oxazole
Molecular Weight (g/mol): 285.08
Molecular Formula: C10H8INO
InChi Key: DDVPBKUHWSZYIP-UHFFFAOYSA-N
SMILES: CC1=C(I)C(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results