accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOCH3OCl

CAS RN 31301-45-8

IUPAC Name: dimethyl-1,2-oxazole-4-carbonyl chloride
Synonyms: dimethyl-1,2-oxazole-4-carbonyl chloride
Molecular Weight (g/mol): 159.57
Molecular Formula: C6H6ClNO2
InChi Key: MPYGFFPGJMGVSW-UHFFFAOYSA-N
SMILES: CC1=C(C(Cl)=O)C(C)=NO1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)