accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNNBr

CAS RN 3138-86-1

IUPAC Name: 2,3-bis(bromomethyl)quinoxaline
Synonyms: 2,3-bis(bromomethyl)quinoxaline
Molecular Weight (g/mol): 316
Molecular Formula: C10H8Br2N2
InChi Key: LHKFFORGJVELPC-UHFFFAOYSA-N
SMILES: BrCC1=C(CBr)N=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results