accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrSBr

CAS RN 3140-92-9

IUPAC Name: 2,4-dibromothiophene
Synonyms: 2,4-dibromothiophene
Molecular Weight (g/mol): 241.93
Molecular Formula: C4H2Br2S
InChi Key: WAQFYSJKIRRXLP-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=CS1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)