accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrSBr

CAS RN 3140-93-0

IUPAC Name: 2,3-dibromothiophene
Synonyms: thiophene, 2,3-dibromo-
Molecular Weight (g/mol): 241.93
Molecular Formula: C4H2Br2S
InChi Key: ATRJNSFQBYKFSM-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C=CS1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)