accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

RuBrBrBr

CAS RN 314075-07-5

IUPAC Name: tribromoruthenium hydrate
Synonyms: tribromoruthenium hydrate
Molecular Weight (g/mol): 358.8
Molecular Formula: Br3H2ORu
InChi Key: PMMMCGISKBNZES-UHFFFAOYSA-K
SMILES: O.Br[Ru](Br)Br
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results