accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3141-26-2

IUPAC Name: 3,4-dibromothiophene
Synonyms: 3,4-dibromothiophene
Molecular Weight (g/mol): 241.93
Molecular Formula: C4H2Br2S
InChi Key: VGKLVWTVCUDISO-UHFFFAOYSA-N
SMILES: BrC1=CSC=C1Br
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results