accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3143-02-0

IUPAC Name: (3-methyloxetan-3-yl)methanol
Synonyms: (3-methyloxetan-3-yl)methanol
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: NLQMSBJFLQPLIJ-UHFFFAOYSA-N
SMILES: CC1(CO)COC1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)