accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31430-18-9

IUPAC Name: methyl N-[6-(thiophene-2-carbonyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: nocodazole
Molecular Weight (g/mol): 301.32
Molecular Formula: C14H11N3O3S
InChi Key: KYRVNWMVYQXFEU-UHFFFAOYSA-N
SMILES: COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CS1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results