accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONHHNCH3O

CAS RN 3148-73-0

IUPAC Name: N'-acetylacetohydrazide
Synonyms: diacetylhydrazine
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
SMILES: CC(=O)NNC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results