accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOOHOH

CAS RN 31519-22-9

IUPAC Name: 1,4-dihydroxynaphthalene-2-carboxylic acid
Synonyms: 1,4-dihydroxy-2-naphthoic acid
Molecular Weight (g/mol): 204.18
Molecular Formula: C11H8O4
InChi Key: VOJUXHHACRXLTD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C2C=CC=CC2=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results