accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3153-37-5

H3COOCl

CAS RN 3153-37-5

IUPAC Name: methyl 4-chlorobutanoate
Synonyms: methyl 4-chlorobutanoate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: ZZUYIRISBMWFMV-UHFFFAOYSA-N
SMILES: COC(=O)CCCCl
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)