accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31555-59-6

CH3SOCl

CAS RN 31555-59-6

IUPAC Name: 5-methylthiophene-2-carbonyl chloride
Synonyms: 5-methylthiophene-2-carbonyl chloride
Molecular Weight (g/mol): 160.62
Molecular Formula: C6H5ClOS
InChi Key: DJCGIWVUKCNCAT-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C(Cl)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)