accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3159-07-7

IUPAC Name: 2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 227.28
Molecular Formula: C13H9NOS
InChi Key: RTERDTBXBYNZIS-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2SC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results