accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH3COSOOCH3CH3

CAS RN 3162-58-1

IUPAC Name: sulfonylideneoxidane; trimethylamine
Synonyms: sulfur trioxide; trimethylamine
Molecular Weight (g/mol): 139.17
Molecular Formula: C3H9NO3S
InChi Key: DXASQZJWWGZNSF-UHFFFAOYSA-N
SMILES: CN(C)C.O=S(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)