CAS RN 3163-76-6
IUPAC Name:
benzene-1,3,5-tricarbaldehyde
Synonyms:
benzene-1,3,5-tricarbaldehyde
Molecular Weight (g/mol):
162.14
Molecular Formula:
C9H6O3
InChi Key:
AEKQNAANFVOBCU-UHFFFAOYSA-N
SMILES:
O=CC1=CC(C=O)=CC(C=O)=C1
1
–
1
of
1
results
1
1,3,5-Triformylbenzene, 98%
CAS: 3163-76-6 Molecular Formula: C9H6O3 Molecular Weight (g/mol): 162.14 MDL Number: MFCD00182470 InChI Key: AEKQNAANFVOBCU-UHFFFAOYSA-N Synonym: 1,3,5-benzenetricarboxaldehyde,1,3,5-triformylbenzene,trimesaldehyde,1,3,5-benzenetricarbaldehyde,benzene-1,3,5-tricarboxaldehyde,trimesic formaldehyde,1,3,5-triformyl benzene,bzoh31,aekqnaanfvobcu-uhfffaoysa PubChem CID: 2747968 IUPAC Name: benzene-1,3,5-tricarbaldehyde SMILES: O=CC1=CC(C=O)=CC(C=O)=C1