CAS RN 317-46-4
IUPAC Name:
2-fluoro-3-nitrobenzoic acid
Synonyms:
2-fluoro-3-nitrobenzoic acid
Molecular Weight (g/mol):
185.11
Molecular Formula:
C7H4FNO4
InChi Key:
WLGUSLGYTNJJFV-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O
1
–
2
of
2
results
1
2-Fluoro-3-nitrobenzoic acid, 97%
CAS: 317-46-4 Molecular Formula: C7H4FNO4 Molecular Weight (g/mol): 185.11 MDL Number: MFCD06797825 InChI Key: WLGUSLGYTNJJFV-UHFFFAOYSA-N Synonym: 2-fluoro-3-nitrobenzoic acid,2-fluoro-3-nitrobenzoicacid,2-fluoro-3-nitro benzoic acid,3-carboxy-2-fluoronitrobenzene,pubchem1398,acmc-1ajf3,ksc494k8n,benzoic acid,2-fluoro-3-nitro PubChem CID: 18672992 IUPAC Name: 2-fluoro-3-nitrobenzoic acid SMILES: OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O
2
2-Fluoro-3-nitrobenzoic Acid, TRC
CAS: 317-46-4 Molecular Formula: C7H4NO4F Molecular Weight (g/mol): 185.11 IUPAC Name: 2-fluoro-3-nitrobenzoic acid SMILES: OC(=O)c1cccc(c1F)[N+](=O)[O-]