accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3171-45-7

CH3NH2NH2H3C

CAS RN 3171-45-7

IUPAC Name: 4,5-dimethylbenzene-1,2-diamine
Synonyms: 4,5-dimethylbenzene-1,2-diamine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: XSZYBMMYQCYIPC-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(N)C=C1C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)