accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31719-76-3

OOO

CAS RN 31719-76-3

IUPAC Name: 4-(phenoxymethyl)benzoate
Synonyms: 4-(phenoxymethyl)benzoate
Molecular Weight (g/mol): 227.24
Molecular Formula: C14H11O3
InChi Key: GRBUVHSYBRTCIB-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(COC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results