CAS RN 317319-15-6
IUPAC Name:
3,5-bis(trifluoromethyl)benzene-1-carbothioamide
Synonyms:
3,5-bis(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol):
273.2
Molecular Formula:
C9H5F6NS
InChi Key:
OWYYZIVORPIBPZ-UHFFFAOYSA-N
SMILES:
NC(=S)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
3,5-Bis(trifluoromethyl)thiobenzamide, 97%
CAS: 317319-15-6 Molecular Formula: C9H5F6NS Molecular Weight (g/mol): 273.20 MDL Number: MFCD00278692 InChI Key: OWYYZIVORPIBPZ-UHFFFAOYSA-N Synonym: 3,5-bis trifluoromethyl thiobenzamide,3,5-di trifluoromethyl benzene-1-carbothioamide,3,5-bis trifluoromethyl benzenecarbothioamide,3,5-bis trifluoromethyl benzothioamide,benzenecarbothioamide,3,5-bis trifluoromethyl,acmc-20ands,maybridge1_003450,3,5-bis-trifluoromethyl-thiobenzamide,3,5-bis trifluoromethyl benzene-1-carbothioamide PubChem CID: 2806924 IUPAC Name: 3,5-bis(trifluoromethyl)benzenecarbothioamide SMILES: NC(=S)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F