accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3179-76-8

IUPAC Name: (3-aminopropyl)diethoxymethylsilane
Synonyms: (3-aminopropyl)diethoxymethylsilane
Molecular Weight (g/mol): 191.35
Molecular Formula: C8H21NO2Si
InChi Key: HXLAEGYMDGUSBD-UHFFFAOYSA-N
SMILES: CCO[Si](C)(CCCN)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results