accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31794-45-3

IUPAC Name: 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride
Synonyms: 3-oxo-2,4-dihydro-1,4-benzoxazine-6-sulfonyl chloride
Molecular Weight (g/mol): 247.65
Molecular Formula: C8H6ClNO4S
InChi Key: CGTCULUUVYBAPX-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(OCC(=O)N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results