accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3182-95-4

IUPAC Name: (2S)-1-hydroxy-3-phenylpropan-2-aminium
Synonyms: (2S)-1-hydroxy-3-phenylpropan-2-aminium
Molecular Weight (g/mol): 152.22
Molecular Formula: C9H14NO
InChi Key: STVVMTBJNDTZBF-VIFPVBQESA-O
SMILES: [NH3+][C@H](CO)CC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)