accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3187-19-7

H3COOHOHOOH3C

CAS RN 3187-19-7

IUPAC Name: 2-hydroxy-4,6-dimethoxybenzoic acid
Synonyms: 2-hydroxy-4,6-dimethoxybenzoic acid
Molecular Weight (g/mol): 198.17
Molecular Formula: C9H10O5
InChi Key: LEIJLTBRQHFMDL-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(C(O)=O)C(OC)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results