accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONOHOH3C(E)

CAS RN 31874-34-7

IUPAC Name: (E)-N-[(2,4-dimethoxyphenyl)methylidene]hydroxylamine
Synonyms: (E)-N-[(2,4-dimethoxyphenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: SFDRVCQSVTYHLU-UXBLZVDNSA-N
SMILES: COC1=CC=C(\C=N\O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results