accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOSH2N

CAS RN 31891-06-2

IUPAC Name: ethyl 2-aminothiophene-3-carboxylate
Synonyms: ethyl 2-aminothiophene-3-carboxylate
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: MKJQYFVTEPGXIE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results