accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 319-03-9

IUPAC Name: 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-fluoro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 166.11
Molecular Formula: C8H3FO3
InChi Key: XVMKZAAFVWXIII-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(=O)OC(=O)C2=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results