CAS RN 319-24-4
IUPAC Name:
methyl 2-amino-5-fluorobenzoate
Synonyms:
methyl 2-amino-5-fluorobenzoate
Molecular Weight (g/mol):
169.16
Molecular Formula:
C8H8FNO2
InChi Key:
PUDDYSBKCDKATP-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(N)C=CC(F)=C1
1
–
1
of
1
results
1
Methyl 2-amino-5-fluorobenzoate, 98%
CAS: 319-24-4 Molecular Formula: C8H8FNO2 Molecular Weight (g/mol): 169.155 MDL Number: MFCD03791308 InChI Key: PUDDYSBKCDKATP-UHFFFAOYSA-N Synonym: 2-amino-5-fluorobenzoic acid methyl ester,methyl2-amino-5-fluorobenzoate,methyl 5-fluoroanthranilate,methyl 2-amino-5-fluoro-benzoate,benzoic acid, 2-amino-5-fluoro-, methyl ester,2-amino-5-fluoro benzoic acid methyl ester,2-amino-5-fluoro-benzoic acid methyl ester,pubchem3558,acmc-209hpn,ksc495g7j PubChem CID: 2783401 IUPAC Name: methyl 2-amino-5-fluorobenzoate SMILES: COC(=O)C1=C(C=CC(=C1)F)N