accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

IBrCl

CAS RN 31928-46-8

IUPAC Name: 2-bromo-1-chloro-4-iodobenzene
Synonyms: 2-bromo-1-chloro-4-iodobenzene
Molecular Weight (g/mol): 317.35
Molecular Formula: C6H3BrClI
InChi Key: KSCPVCBJIZDZER-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results